Product Details

SNP ID
rs150108952
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:197084342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTTACTAATAAGGAATGCCAAG[C/T]GTATCCATCACCATTTGAATAGCTT
Phenotype
MIM: 605481
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASPM PubMed Links

Gene Details

Gene
ASPM
Gene Name
abnormal spindle microtubule assembly
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206846.1 10673 Silent Mutation ACA,ACG T1887T NP_001193775.1
NM_018136.4 10673 Silent Mutation ACA,ACG T3472T NP_060606.3

View Full Product Details