Product Details

SNP ID
rs150159034
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:100151339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTCTGTCTCCTAATTGTCTTTA[C/T]ATATTCTCTGCTTTCCAAAACTAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC39 PubMed Links

Gene Details

Gene
LRRC39
Gene Name
leucine rich repeat containing 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256385.1 Intron NP_001243314.1
NM_001256386.1 Intron NP_001243315.1
NM_001256387.1 Intron NP_001243316.1
NM_144620.3 Intron NP_653221.1
Gene
TRMT13
Gene Name
tRNA methyltransferase 13 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019083.2 Intron NP_061956.2
XM_005270945.3 Intron XP_005271002.1
XM_005270946.2 Intron XP_005271003.1
XM_017001504.1 Intron XP_016856993.1
XM_017001505.1 Intron XP_016856994.1

View Full Product Details