Product Details

SNP ID
rs150239478
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:111444063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGCTATCTCTTACCTCGCTGCA[C/T]GAAAGAAACACAGAGTCCTTGTGAG
Phenotype
MIM: 603270 MIM: 611734
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5F1 PubMed Links

Gene Details

Gene
ATP5F1
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1
There are no transcripts associated with this gene.

Gene
WDR77
Gene Name
WD repeat domain 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317062.1 694 Silent Mutation TCA,TCG S185S NP_001303991.1
NM_001317063.1 694 Silent Mutation TCA,TCG S185S NP_001303992.1
NM_001317064.1 694 Silent Mutation TCA,TCG S121S NP_001303993.1
NM_024102.3 694 Silent Mutation TCA,TCG S185S NP_077007.1

View Full Product Details