Product Details

SNP ID
rs150299630
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:112917305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACAATACGGAGCTGAGCCACCC[A/G]AAGGCAAATCCAAAGAATCCCGCAT
Phenotype
MIM: 600682
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKR7A2P1 PubMed Links

Gene Details

Gene
AKR7A2P1
Gene Name
aldo-keto reductase family 7 member A2 pseudogene 1
There are no transcripts associated with this gene.

Gene
SLC16A1
Gene Name
solute carrier family 16 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166496.1 1464 Silent Mutation TTC,TTT F367F NP_001159968.1
NM_003051.3 1464 Silent Mutation TTC,TTT F367F NP_003042.3
XM_011542026.2 1464 Intron XP_011540328.1
XM_011542027.2 1464 Intron XP_011540329.1

View Full Product Details