Product Details

SNP ID
rs150320192
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:70260398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATTGAAGATGACTATACGATTGT[C/T]CCCCTACCAATTTAAACAGTTTCAA
Phenotype
MIM: 615125 MIM: 602010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD13C PubMed Links

Gene Details

Gene
ANKRD13C
Gene Name
ankyrin repeat domain 13C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030816.4 3240 UTR 3 NP_110443.3
XM_005271234.3 3240 UTR 3 XP_005271291.1
XM_005271235.3 3240 Intron XP_005271292.1
XM_006710929.3 3240 Intron XP_006710992.1
XM_017002413.1 3240 UTR 3 XP_016857902.1
XM_017002414.1 3240 Intron XP_016857903.1
XM_017002415.1 3240 Intron XP_016857904.1
XM_017002416.1 3240 UTR 3 XP_016857905.1
Gene
SRSF11
Gene Name
serine and arginine rich splicing factor 11
There are no transcripts associated with this gene.

View Full Product Details