Product Details

SNP ID
rs150366466
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:118884864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCATGCTGTGCTCCATCCCTGA[C/T]GGCAGGTACTGCCTCTCTCCAAAGG
Phenotype
MIM: 604127
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBX15 PubMed Links

Gene Details

Gene
TBX15
Gene Name
T-box 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152380.2 2130 Silent Mutation CCA,CCG P453P NP_689593.2
XM_005271161.3 2130 Silent Mutation CCA,CCG P592P XP_005271218.1
XM_005271162.1 2130 Silent Mutation CCA,CCG P559P XP_005271219.1
XM_011542052.2 2130 Silent Mutation CCA,CCG P277P XP_011540354.1

View Full Product Details