Product Details
- SNP ID
-
rs150404763
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:27613262 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTGGTGATGAGCTCAGTGAGCAG[G/T]ATCCCAAAGGACCACACGTCTGACT
- Phenotype
-
MIM: 615790
MIM: 164940
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AHDC1
PubMed Links
Gene Details
- Gene
- AHDC1
- Gene Name
- AT-hook DNA binding motif containing 1
There are no transcripts associated with this gene.
- Gene
- FGR
- Gene Name
- FGR proto-oncogene, Src family tyrosine kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001042729.1 |
1566 |
Silent Mutation |
ATA,ATC |
I446I |
NP_001036194.1 |
NM_001042747.1 |
1566 |
Silent Mutation |
ATA,ATC |
I446I |
NP_001036212.1 |
NM_005248.2 |
1566 |
Silent Mutation |
ATA,ATC |
I446I |
NP_005239.1 |
XM_006710452.2 |
1566 |
Silent Mutation |
ATA,ATC |
I446I |
XP_006710515.1 |
XM_011541010.1 |
1566 |
Silent Mutation |
ATA,ATC |
I446I |
XP_011539312.1 |
XM_011541011.2 |
1566 |
Silent Mutation |
ATA,ATC |
I377I |
XP_011539313.1 |
XM_011541012.1 |
1566 |
Missense Mutation |
TAC,TCC |
Y395S |
XP_011539314.1 |
XM_011541013.2 |
1566 |
Intron |
|
|
XP_011539315.1 |
XM_011541014.2 |
1566 |
Silent Mutation |
ATA,ATC |
I242I |
XP_011539316.1 |
XM_017000673.1 |
1566 |
Silent Mutation |
ATA,ATC |
I242I |
XP_016856162.1 |
XM_017000674.1 |
1566 |
Silent Mutation |
ATA,ATC |
I242I |
XP_016856163.1 |
View Full Product Details