Product Details

SNP ID
rs150417585
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:109715232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAAACTTGAAGGACTTCATCTCC[C/T]GCTTTGAGGTGATGCCCCCATCCTC
Phenotype
MIM: 138385
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTM5 PubMed Links

Gene Details

Gene
GSTM5
Gene Name
glutathione S-transferase mu 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000851.3 656 Missense Mutation CGC,TGC R187C NP_000842.2
XM_005270784.4 656 Missense Mutation CGC,TGC R187C XP_005270841.1
XM_005270785.4 656 Missense Mutation CGC,TGC R83C XP_005270842.1

View Full Product Details