Product Details

SNP ID
rs150493209
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:101239216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGGAAAACCAAGAAATTCCAC[A/C]GACCCATGTACTATTTTATTGGCAA
Phenotype
MIM: 601974
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928370 PubMed Links

Gene Details

Gene
LOC101928370
Gene Name
uncharacterized LOC101928370
There are no transcripts associated with this gene.

Gene
S1PR1
Gene Name
sphingosine-1-phosphate receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320730.1 638 Silent Mutation AGA,CGA R78R NP_001307659.1
NM_001400.4 638 Silent Mutation AGA,CGA R78R NP_001391.2

View Full Product Details