Product Details

SNP ID
rs150532144
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:23848774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTGGAAGATGGGAACAATCAG[A/T]CCATCTTTAGTTGGTCCAATGTTCA
Phenotype
MIM: 612280
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FUCA1 PubMed Links

Gene Details

Gene
FUCA1
Gene Name
fucosidase, alpha-L- 1, tissue
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000147.4 703 Silent Mutation GGA,GGT G345G NP_000138.2
XM_005245821.2 703 Silent Mutation GGA,GGT G220G XP_005245878.1
XM_011541167.2 703 Silent Mutation GGA,GGT G134G XP_011539469.1
XM_017000905.1 703 Silent Mutation GGA,GGT G244G XP_016856394.1

View Full Product Details