Product Details

SNP ID
rs150540703
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:158181557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCGTGGCTGGGGGAGCTGCAGA[C/T]GCACAGCTGGAGCAACGACTCGGAC
Phenotype
MIM: 188410
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD1D PubMed Links

Gene Details

Gene
CD1D
Gene Name
CD1d molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319145.1 342 Missense Mutation ACG,ATG T55M NP_001306074.1
NM_001766.3 342 Missense Mutation ACG,ATG T55M NP_001757.1
XM_005245583.1 342 UTR 5 XP_005245640.1
XM_006711621.1 342 UTR 5 XP_006711684.1
XM_011510125.1 342 Missense Mutation ACG,ATG T55M XP_011508427.1
XM_011510127.1 342 UTR 5 XP_011508429.1

View Full Product Details