Product Details

SNP ID
rs150557296
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:145825886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAGAACCGGCAGCCAGCCACTAA[T/G]AGTAATTGCCCATGACACAGGAACA
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
POLR3C PubMed Links

Gene Details

Gene
POLR3C
Gene Name
RNA polymerase III subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303456.1 233 Missense Mutation NP_001290385.1
NM_006468.7 233 Missense Mutation NP_006459.3
XM_005272927.3 233 UTR 5 XP_005272984.1
XM_005272928.3 233 UTR 5 XP_005272985.1
XM_011509078.2 233 Missense Mutation XP_011507380.1
Gene
RNF115
Gene Name
ring finger protein 115
There are no transcripts associated with this gene.

View Full Product Details