Product Details

SNP ID
rs150640693
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:28148421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTTCCCAGCCCATGTGCCTGTT[C/T]GTATGGAAGCCCCTGGACATCGGGG
Phenotype
MIM: 173393
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTAFR PubMed Links

Gene Details

Gene
PTAFR
Gene Name
platelet activating factor receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000952.4 2936 UTR 3 NP_000943.1
NM_001164721.1 2936 UTR 3 NP_001158193.1
NM_001164722.2 2936 UTR 3 NP_001158194.1
NM_001164723.2 2936 UTR 3 NP_001158195.1

View Full Product Details