Product Details

SNP ID
rs150734016
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:25225014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAATACTTACTGTTTTTCCAGA[C/T]CTATGACCATCCTGTCAATTTCCTC
Phenotype
MIM: 607090
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SYF2 PubMed Links

Gene Details

Gene
SYF2
Gene Name
SYF2 pre-mRNA splicing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015484.4 483 Missense Mutation GAT,GGT D185G NP_056299.1
NM_207170.3 483 Missense Mutation GAT,GGT D143G NP_997053.1

View Full Product Details