Product Details
- SNP ID
-
rs150740989
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:159313794 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTCCTTGTTCTTCAGGCTGTACA[C/G]AACAGGGTTCAGTAGGGGAGTGATG
- Phenotype
-
MIM: 147140
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FCER1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs58037016] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FCER1A
- Gene Name
- Fc fragment of IgE receptor Ia
There are no transcripts associated with this gene.
- Gene
- OR10J3
- Gene Name
- olfactory receptor family 10 subfamily J member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004467.1 |
866 |
Missense Mutation |
TCT,TGT |
S289C |
NP_001004467.1 |
View Full Product Details