Product Details
- SNP ID
-
rs150759144
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:3739024 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGGCATCTACCCATTGAGCATCA[A/G]GACATCCCAGGTTCCTCCTGAAAAC
- Phenotype
-
MIM: 601990
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TP73
PubMed Links
Gene Details
- Gene
- TP73
- Gene Name
- tumor protein p73
There are no transcripts associated with this gene.
- Gene
- TP73-AS1
- Gene Name
- TP73 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details