Product Details

SNP ID
rs150771597
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:111454237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTGCAGGCAACAAGGACCTTTC[A/G]TACAGGGCAGCCACACCTTGTCCCT
Phenotype
MIM: 603270 MIM: 611734
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP5F1 PubMed Links

Gene Details

Gene
ATP5F1
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001688.4 525 Missense Mutation CAT,CGT H35R NP_001679.2
Gene
C1orf162
Gene Name
chromosome 1 open reading frame 162
There are no transcripts associated with this gene.

Gene
WDR77
Gene Name
WD repeat domain 77
There are no transcripts associated with this gene.

View Full Product Details