Product Details
- SNP ID
-
rs150888098
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.1:3754148 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCAAGGATGCCGCCAGCCTCACC[A/G]GCTTCATGTCCAAAATGGAACTTCG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC27
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149705892] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC27
- Gene Name
- coiled-coil domain containing 27
- Gene
- TP73-AS1
- Gene Name
- TP73 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details