Product Details

SNP ID
rs150928044
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:206637697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTCATGATGATAGATGAAACCA[A/G]ATGTCCCCCCTGTTCAAATGTACTC
Phenotype
MIM: 603497
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYRK3 PubMed Links

Gene Details

Gene
DYRK3
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004023.1 387 Missense Mutation AAA,AGA K22R NP_001004023.1
NM_003582.2 387 Missense Mutation AAA,AGA K42R NP_003573.2
XM_005273315.4 387 Missense Mutation AAA,AGA K22R XP_005273372.1
XM_011510061.2 387 Missense Mutation AAA,AGA K7R XP_011508363.1

View Full Product Details