Product Details

SNP ID
rs151027751
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:11109695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTGTGGTCCCCGTTTTCTTATG[C/G]GCTGGCTCTCCAAGTTCCACACCGT
Phenotype
MIM: 605960 MIM: 601231
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EXOSC10 PubMed Links

Gene Details

Gene
EXOSC10
Gene Name
exosome component 10
There are no transcripts associated with this gene.

Gene
LOC105376736
Gene Name
uncharacterized LOC105376736
There are no transcripts associated with this gene.

Gene
MTOR
Gene Name
mechanistic target of rapamycin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004958.3 6985 Silent Mutation GCC,GCG A2467A NP_004949.1
XM_005263438.2 6985 Silent Mutation GCC,GCG A2467A XP_005263495.1
XM_011541166.2 6985 Intron XP_011539468.1
XM_017000900.1 6985 Silent Mutation GCC,GCG A2240A XP_016856389.1
XM_017000901.1 6985 Silent Mutation GCC,GCG A2051A XP_016856390.1
XM_017000902.1 6985 Intron XP_016856391.1

View Full Product Details