Product Details

SNP ID
rs151055866
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:62238374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGAGCGATGGACAAAGCTGTG[C/T]GGCCAGCCTACAGGAGAAAAAGGAA
Phenotype
MIM: 614612
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KANK4 PubMed Links

Gene Details

Gene
KANK4
Gene Name
KN motif and ankyrin repeat domains 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320269.1 1522 Missense Mutation NP_001307198.1
NM_181712.4 1522 Missense Mutation NP_859063.3
XM_011540843.2 1522 Missense Mutation XP_011539145.1
XM_011540845.2 1522 Missense Mutation XP_011539147.1
XM_017000483.1 1522 Missense Mutation XP_016855972.1
XM_017000484.1 1522 Missense Mutation XP_016855973.1
XM_017000485.1 1522 Missense Mutation XP_016855974.1

View Full Product Details