Product Details

SNP ID
rs151087109
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:212835988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGTATCAGTAAAGGACTGATA[A/G]GCATCCGGGAAGTGCCTCTTTTGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPATA45 PubMed Links

Gene Details

Gene
SPATA45
Gene Name
spermatogenesis associated 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024601.2 270 Silent Mutation GCC,GCT A54A NP_001019772.1

View Full Product Details