Product Details

SNP ID
rs151104998
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:151986102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTAAGCTTTTCAACACTTACTTC[C/T]AAAAATCCAGGGAACTCCTTTTCCA
Phenotype
MIM: 114085
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S100A10 PubMed Links

Gene Details

Gene
S100A10
Gene Name
S100 calcium binding protein A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002966.2 637 Silent Mutation TTA,TTG L43L NP_002957.1

View Full Product Details