Product Details

SNP ID
rs151227410
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:32779484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCTCACCAGGAGCAGAGCCTGC[C/T]GGAGGGTCCAGAGGTTCAACCTGGC
Phenotype
MIM: 603623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1522 PubMed Links

Gene Details

Gene
KIAA1522
Gene Name
KIAA1522
There are no transcripts associated with this gene.

Gene
YARS
Gene Name
tyrosyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003680.3 1161 Silent Mutation CCA,CCG P458P NP_003671.1
XM_011542347.1 1161 Silent Mutation CCA,CCG P248P XP_011540649.1
XM_011542348.1 1161 Silent Mutation CCA,CCG P248P XP_011540650.1
XM_017002651.1 1161 Silent Mutation CCA,CCG P248P XP_016858140.1

View Full Product Details