Product Details

SNP ID
rs151327006
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:162374146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAGGAGAGCCTGTCTCCCAGCC[G/T]GGCCTTCTGCTCCTTGCTCCAGAGG
Phenotype
MIM: 605551
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C1orf111 PubMed Links

Gene Details

Gene
C1orf111
Gene Name
chromosome 1 open reading frame 111
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182581.3 752 Silent Mutation NP_872387.2
XM_005245103.3 752 Missense Mutation XP_005245160.1
XM_011509443.2 752 Missense Mutation XP_011507745.1
XM_011509444.1 752 Missense Mutation XP_011507746.1
XM_011509445.2 752 Intron XP_011507747.1
Gene
C1orf226
Gene Name
chromosome 1 open reading frame 226
There are no transcripts associated with this gene.

Gene
NOS1AP
Gene Name
nitric oxide synthase 1 adaptor protein
There are no transcripts associated with this gene.

View Full Product Details