Product Details

SNP ID
rs151330882
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:158717518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATGATGGTCACAGCATGAATCA[C/T]ATCCTCAATCAGAATCATGGACGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6K3 PubMed Links

Gene Details

Gene
OR6K3
Gene Name
olfactory receptor family 6 subfamily K member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005327.2 598 Missense Mutation ATG,GTG M200V NP_001005327.2

View Full Product Details