Product Details
- SNP ID
-
rs137965181
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:38213445 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTGCCACCCCTCTTCGTTCTGGG[C/T]GCGCTGAGTGAGCAGCTGGAGCCTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1755
PubMed Links
Gene Details
- Gene
- KIAA1755
- Gene Name
- KIAA1755
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001029864.1 |
3472 |
Missense Mutation |
CAC,CGC |
H1067R |
NP_001025035.1 |
View Full Product Details