Product Details
- SNP ID
-
rs138150427
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:188117 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAGCATTTTTTGAGTCTTGCCCC[A/G]TCTGTGCATAGGAAACAACATTGAA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB128
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35089290] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DEFB128
- Gene Name
- defensin beta 128
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001037732.2 |
109 |
Silent Mutation |
GAC,GAT |
D17D |
NP_001032821.1 |
View Full Product Details