Product Details

SNP ID
rs138379271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58460899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGCAGCGATGGCCTGTGCTGC[A/G]GTGGCCTGGAGAAGTCGGGGGCCTC
Phenotype
MIM: 605704
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APCDD1L PubMed Links

Gene Details

Gene
APCDD1L
Gene Name
APC down-regulated 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304787.1 1628 Missense Mutation CCG,CTG P477L NP_001291716.1
NM_153360.2 1628 Missense Mutation CCG,CTG P466L NP_699191.1
Gene
VAPB
Gene Name
VAMP associated protein B and C
There are no transcripts associated with this gene.

View Full Product Details