Product Details

SNP ID
rs138408753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63236317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAGGAGGAGGGCGCCGGGGCCA[C/T]CTTGTCCAGGGGGCCTGCCTTCCCC
Phenotype
MIM: 605737 MIM: 612873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BIRC7 PubMed Links

Gene Details

Gene
BIRC7
Gene Name
baculoviral IAP repeat containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022161.3 435 Missense Mutation ACC,ATC T74I NP_071444.1
NM_139317.2 435 Missense Mutation ACC,ATC T74I NP_647478.1
Gene
LOC107985425
Gene Name
uncharacterized LOC107985425
There are no transcripts associated with this gene.

Gene
MIR3196
Gene Name
microRNA 3196
There are no transcripts associated with this gene.

Gene
NKAIN4
Gene Name
Na+/K+ transporting ATPase interacting 4
There are no transcripts associated with this gene.

View Full Product Details