Product Details

SNP ID
rs138509176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35616014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGGGGGTCAGGATGCGGCGAA[A/G]CTCCCGCCCGGGCTCGGCCTCGTCC
Phenotype
MIM: 603038
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FER1L4 PubMed Links

Gene Details

Gene
FER1L4
Gene Name
fer-1 like family member 4, pseudogene
There are no transcripts associated with this gene.

Gene
SPAG4
Gene Name
sperm associated antigen 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317931.1 131 UTR 5 NP_001304860.1
NM_003116.2 131 Missense Mutation AAC,AGC N4S NP_003107.1
XM_005260520.4 131 Intron XP_005260577.1
XM_011529009.2 131 Missense Mutation AAC,AGC N4S XP_011527311.1

View Full Product Details