Product Details

SNP ID
rs138741351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:41048221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACATGAATTTTTGTAGGTGGCT[C/T]GTATTTGCTATTAATATTGCTCAGA
Phenotype
MIM: 126420
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOP1 PubMed Links

Gene Details

Gene
TOP1
Gene Name
topoisomerase (DNA) I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003286.2 Intron NP_003277.1

View Full Product Details