Product Details

SNP ID
rs138989733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:40688624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTCGAGGTGTGTCTTCTGTTCG[G/T]TCGGGCTGAAGCTGGGCGACGAGGG
Phenotype
MIM: 608968
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAFB PubMed Links

Gene Details

Gene
MAFB
Gene Name
MAF bZIP transcription factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005461.4 617 Missense Mutation AAC,ACC N76T NP_005452.2

View Full Product Details