Product Details

SNP ID
rs139280028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50128031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGACGTGGTGGATGCGATGGTGT[C/T]TACGTGGCAGGATGACATGCCAGTC
Phenotype
MIM: 610994
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM189 PubMed Links

Gene Details

Gene
TMEM189
Gene Name
transmembrane protein 189
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162505.1 796 Missense Mutation AAA,AGA K209R NP_001155977.1
NM_199129.2 796 Missense Mutation AAA,AGA K212R NP_954580.1
Gene
TMEM189-UBE2V1
Gene Name
TMEM189-UBE2V1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199203.2 796 Missense Mutation AAA,AGA K212R NP_954673.1

View Full Product Details