Product Details

SNP ID
rs139314100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62709709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGCCTGAGTGTGGAGCGCTAC[A/C]TGGCCATCTGCCACCCCTTCAAGGC
Phenotype
MIM: 162651
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC107987285 PubMed Links

Gene Details

Gene
LOC107987285
Gene Name
uncharacterized LOC107987285
There are no transcripts associated with this gene.

Gene
NTSR1
Gene Name
neurotensin receptor 1 (high affinity)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002531.2 873 Missense Mutation ATG,CTG M168L NP_002522.2
XM_011528827.2 873 Missense Mutation ATG,CTG M168L XP_011527129.1

View Full Product Details