Product Details

SNP ID
rs139413270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35434256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGCTGGGTCGCTTGCCCTGGC[A/G]AGTGGCCAGTGGGGCCCGCCGTTTT
Phenotype
MIM: 601146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GDF5 PubMed Links

Gene Details

Gene
GDF5
Gene Name
growth differentiation factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000557.4 1421 Missense Mutation CGC,TGC R387C NP_000548.2
NM_001319138.1 1421 Missense Mutation CGC,TGC R387C NP_001306067.1
XM_011529075.2 1421 Missense Mutation CGC,TGC R387C XP_011527377.1

View Full Product Details