Product Details

SNP ID
rs139481302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:6075332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGACCAGTTATAATCTTAAAGA[A/G]CTGACTAGGTTCTAAAATAATAGAA
Phenotype
MIM: 607900
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FERMT1 PubMed Links

Gene Details

Gene
FERMT1
Gene Name
fermitin family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017671.4 4664 UTR 3 NP_060141.3

View Full Product Details