Product Details
- SNP ID
-
rs139565347
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:64106113 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTCCAGAGCGGAGCCGCACGGCC[C/T]GCAGCCTGCGCAGGAGGTCTGTGTA
- Phenotype
-
MIM: 600731
MIM: 602548
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NPBWR2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4809400] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NPBWR2
- Gene Name
- neuropeptides B/W receptor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005286.2 |
719 |
Missense Mutation |
CAG,CGG |
Q240R |
NP_005277.2 |
- Gene
- OPRL1
- Gene Name
- opioid related nociceptin receptor 1
There are no transcripts associated with this gene.
View Full Product Details