Product Details

SNP ID
rs139609295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31640033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCACAGAACTCAACGCTGAGGA[A/G]CAGGCCCTGAAGGAGAAGGAGAAGG
Phenotype
MIM: 607976
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX4I2 PubMed Links

Gene Details

Gene
COX4I2
Gene Name
cytochrome c oxidase subunit 4I2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032609.2 258 Silent Mutation GAA,GAG E61E NP_115998.2
XM_005260579.4 258 Silent Mutation GAA,GAG E66E XP_005260636.1
XM_005260580.4 258 Silent Mutation GAA,GAG E66E XP_005260637.1
XM_005260581.3 258 Silent Mutation GAA,GAG E61E XP_005260638.1

View Full Product Details