Product Details

SNP ID
rs139630946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45948991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTGATGACGTCGTACTCGATGC[C/T]CTGGTGCTGCAGCTGTTGAATGTGT
Phenotype
MIM: 610827
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCIF1 PubMed Links

Gene Details

Gene
PCIF1
Gene Name
PDX1 C-terminal inhibiting factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022104.3 3885 Intron NP_071387.1
XM_011528980.2 3885 Intron XP_011527282.1
XM_011528981.2 3885 Intron XP_011527283.1
XM_017028013.1 3885 Intron XP_016883502.1
XM_017028014.1 3885 Intron XP_016883503.1
Gene
ZNF335
Gene Name
zinc finger protein 335
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022095.3 3885 Missense Mutation AGC,GGC S1331G NP_071378.1
XM_005260504.4 3885 Missense Mutation AGC,GGC S1330G XP_005260561.1
XM_011528979.2 3885 Intron XP_011527281.1
XM_017028012.1 3885 Missense Mutation AGC,GGC S1155G XP_016883501.1

View Full Product Details