Product Details

SNP ID
rs139855814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63642166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGCCTCGCGGATCTCCTTGCTG[A/C]GCTCCATCTTGTAGTTGAGCTTCTC
Phenotype
MIM: 608362
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
STMN3 PubMed Links

Gene Details

Gene
STMN3
Gene Name
stathmin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276310.1 690 Missense Mutation CGC,CTC R131L NP_001263239.1
NM_015894.3 690 Missense Mutation CGC,CTC R142L NP_056978.2

View Full Product Details