Product Details

SNP ID
rs140037064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63944627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCTTGACAAACTTGTTGTACTG[C/T]TTGATGACACCCTCGATGTCCCGGC
Phenotype
MIM: 611203 MIM: 610866
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC5 PubMed Links

Gene Details

Gene
DNAJC5
Gene Name
DnaJ heat shock protein family (Hsp40) member C5
There are no transcripts associated with this gene.

Gene
MIR1914
Gene Name
microRNA 1914
There are no transcripts associated with this gene.

Gene
MIR647
Gene Name
microRNA 647
There are no transcripts associated with this gene.

Gene
UCKL1
Gene Name
uridine-cytidine kinase 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193379.1 1037 Silent Mutation NP_001180308.1
NM_017859.3 1037 Silent Mutation NP_060329.2
XM_005260216.2 1037 Missense Mutation XP_005260273.1
XM_006723806.2 1037 Missense Mutation XP_006723869.1
XM_006723807.2 1037 Missense Mutation XP_006723870.1
XM_006723808.1 1037 Missense Mutation XP_006723871.1
XM_006723809.1 1037 Missense Mutation XP_006723872.1
XM_006723810.1 1037 Missense Mutation XP_006723873.1
XM_006723811.3 1037 Missense Mutation XP_006723874.1
XM_011528868.2 1037 Missense Mutation XP_011527170.1
XM_011528869.1 1037 Missense Mutation XP_011527171.1
XM_011528870.1 1037 Silent Mutation XP_011527172.1
XM_011528871.1 1037 Missense Mutation XP_011527173.1
XM_011528872.1 1037 Missense Mutation XP_011527174.1
XM_011528873.2 1037 Missense Mutation XP_011527175.1
XM_017027894.1 1037 Missense Mutation XP_016883383.1
XM_017027895.1 1037 Missense Mutation XP_016883384.1
XM_017027896.1 1037 Missense Mutation XP_016883385.1
XM_017027897.1 1037 Missense Mutation XP_016883386.1
XM_017027898.1 1037 Missense Mutation XP_016883387.1
XM_017027899.1 1037 Missense Mutation XP_016883388.1
XM_017027900.1 1037 Missense Mutation XP_016883389.1
XM_017027901.1 1037 Missense Mutation XP_016883390.1
XM_017027902.1 1037 Missense Mutation XP_016883391.1
XM_017027903.1 1037 Missense Mutation XP_016883392.1
XM_017027904.1 1037 Missense Mutation XP_016883393.1
XM_017027905.1 1037 Missense Mutation XP_016883394.1
XM_017027906.1 1037 Missense Mutation XP_016883395.1
XM_017027907.1 1037 Missense Mutation XP_016883396.1
XM_017027908.1 1037 Missense Mutation XP_016883397.1
Gene
UCKL1-AS1
Gene Name
UCKL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details