Product Details

SNP ID
rs140109710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:8139851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTATAATACAAAATCTGGACTTC[C/T]AAAGCTACATTTACCAGACTCCTTT
Phenotype
MIM: 607120
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLCB1 PubMed Links

Gene Details

Gene
PLCB1
Gene Name
phospholipase C beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015192.3 Intron NP_056007.1
NM_182734.2 Intron NP_877398.1
XM_011529199.2 Intron XP_011527501.1
XM_011529200.2 Intron XP_011527502.1
XM_011529201.2 Intron XP_011527503.1
XM_011529203.2 Intron XP_011527505.1
XM_017027752.1 Intron XP_016883241.1
XM_017027753.1 Intron XP_016883242.1

View Full Product Details