Product Details

SNP ID
rs140273478
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:64073226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAATAAGACATCCTATTTTTGGG[A/G]GGTTAGGGATAGAGATAAATATGAA
Phenotype
MIM: 602548 MIM: 605071 MIM: 604784
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MIR6813 PubMed Links

Gene Details

Gene
MIR6813
Gene Name
microRNA 6813
There are no transcripts associated with this gene.

Gene
OPRL1
Gene Name
opioid related nociceptin receptor 1
There are no transcripts associated with this gene.

Gene
RGS19
Gene Name
regulator of G-protein signaling 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039467.1 1548 UTR 3 NP_001034556.1
NM_005873.2 1548 UTR 3 NP_005864.1
XM_005260182.3 1548 UTR 3 XP_005260239.1
XM_005260183.2 1548 UTR 3 XP_005260240.1
XM_011528484.1 1548 UTR 3 XP_011526786.1
XM_011528485.2 1548 UTR 3 XP_011526787.1
XM_011528486.2 1548 UTR 3 XP_011526788.1
Gene
TCEA2
Gene Name
transcription elongation factor A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003195.4 1548 Intron NP_003186.1
NM_198723.1 1548 Intron NP_942016.1
XM_005260229.3 1548 Intron XP_005260286.1
XM_011529022.1 1548 Intron XP_011527324.1
XM_011529023.1 1548 Intron XP_011527325.1
XM_011529027.2 1548 Intron XP_011527329.1
XM_017028036.1 1548 Intron XP_016883525.1
XM_017028037.1 1548 Intron XP_016883526.1

View Full Product Details