Product Details

SNP ID
rs140317494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62464942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGCAAAGTCCTCAGGCTCGAAC[C/T]TGAACTCCAAGTGGCCGGGGGCAAG
Phenotype
MIM: 611496
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GATA5 PubMed Links

Gene Details

Gene
GATA5
Gene Name
GATA binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080473.4 1150 Missense Mutation AAG,AGG K363R NP_536721.1
XM_006723699.3 1150 Missense Mutation AAG,AGG K363R XP_006723762.1

View Full Product Details