Product Details

SNP ID
rs140478149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:64106030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGCGTCCAGCAGAGGAGGCACA[C/T]GGCCAGCACGACGAGGACCAGGACG
Phenotype
MIM: 600731 MIM: 602548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPBWR2 PubMed Links

Gene Details

Gene
NPBWR2
Gene Name
neuropeptides B/W receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005286.2 802 Missense Mutation ATG,GTG M268V NP_005277.2
Gene
OPRL1
Gene Name
opioid related nociceptin receptor 1
There are no transcripts associated with this gene.

View Full Product Details