Product Details

SNP ID
rs140729217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:13715046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGTTCTTTCCGTTCTCTTTGC[C/T]GGGCCTTCTCCTCAAGGATTTTTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESF1 PubMed Links

Gene Details

Gene
ESF1
Gene Name
ESF1 nucleolar pre-rRNA processing protein homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276380.1 3944 Missense Mutation CAG,CGG Q795R NP_001263309.1
NM_016649.3 3944 Missense Mutation CAG,CGG Q795R NP_057733.2
XM_017027874.1 3944 Missense Mutation CAG,CGG Q795R XP_016883363.1

View Full Product Details