Product Details
- SNP ID
-
rs140875990
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:38213196 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGGTGGTGGCAAGCAAATGCTCG[C/T]GGGCAGGGTCAGGCAGCTTGTGGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1755
PubMed Links
Gene Details
- Gene
- KIAA1755
- Gene Name
- KIAA1755
View Full Product Details