Product Details

SNP ID
rs140885197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44306763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACAAGGTGCCAAACACCTTCTG[A/G]GACAAGTTGTGGAAATAAACAGCTG
Phenotype
MIM: 612029
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FITM2 PubMed Links

Gene Details

Gene
FITM2
Gene Name
fat storage inducing transmembrane protein 2
There are no transcripts associated with this gene.

View Full Product Details