Product Details

SNP ID
rs141173032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:40688137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGGTCCGCCGCTTCTGCTTCAG[A/G]CGGATCACCTCGTCCTTGGTGAAGC
Phenotype
MIM: 608968
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAFB PubMed Links

Gene Details

Gene
MAFB
Gene Name
MAF bZIP transcription factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005461.4 1104 Silent Mutation CGC,CGT R238R NP_005452.2

View Full Product Details